4-Methoxycarbonyl-2-nitrophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Methoxycarbonyl)-2-nitrophenylboronic acid, 4-(Methoxycarbonyl)-2-nitrobenzeneboronic acid, 4-Methoxycarbonyl-2-nitrobenzeneboronic acid

A343749-10G A343749-1G A343749-25G A343749-5G A343749-100G
77239-924EA 42 USD
77239-924 77239-926 77239-928 77746-672 77746-671
4-Methoxycarbonyl-2-nitrophenylboronic acid 97%
4-Methoxycarbonyl-2-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₆
MDL Number: MFCD01632202
CAS Number: 85107-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR