4-Chloro-1,3-difluorobenzene 98%

Supplier: Ambeed
Danger

Synonyms: 1-Chloro-2,4-difluorobenzene

A343531-5G A343531-25G
77239-922EA 7.21 USD
77239-922 77550-992
4-Chloro-1,3-difluorobenzene 98%
4-Chloro-1,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClF₂
MW: 148.54 g/mol
MDL Number: MFCD00042569
CAS Number: 1435-44-5
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR