3-Fluoro-4-isopropoxyphenylboronicacid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-4-isopropoxyphenylboronic acid

A343415-1G A343415-250MG A343415-25G A343415-5G
77278-018EA 43.26 USD
77278-018 77278-020 77278-022 77278-024
3-Fluoro-4-isopropoxyphenylboronicacid 97%
3-Fluoro-4-isopropoxyphenylboronicacid

Precaución: for Research Use Only

Formula: C₉H₁₂BFO₃
MW: 198.00 g/mol
MDL Number: MFCD05865163
CAS Number: 480438-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR