BMS-3 98%

Supplier: Ambeed
Warning

Synonyms: N-(5-(1-(2,6-Dichlorophenyl)-3-(difluoromethyl)-1H-pyrazol-5-yl)thiazol-2-yl)cyclopropanecarboxamide

A343340-100MG A343340-10MG A343340-1MG A343340-50MG A343340-5MG
77239-912EA 344.02 USD
77239-912 77239-914 77239-916 77239-918 77239-920
BMS-3 98%
BMS-3

Precaución: for Research Use Only

Formula: C₁₇H₁₂Cl₂F₂N₄OS
MW: 429.3 g/mol
MDL Number: MFCD17019326
CAS Number: 1338247-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR