(11aS)-1,11-Bis(diphenylphosphino)dibenzo[d,f][1,3]dioxepine 98%

Supplier: Ambeed
Warning

A342902-100MG A342902-1G A342902-250MG
77263-792EA 103 USD
77263-792 77263-794 77263-796
(11aS)-1,11-Bis(diphenylphosphino)dibenzo[d,f][1,3]dioxepine 98%
(11aS)-1,11-Bis(diphenylphosphino)dibenzo[d,f][1,3]dioxepine

Precaución: for Research Use Only

Formula: C₃₇H₂₈O₂P₂
MW: 566.56 g/mol
CAS Number: 486429-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR