2-Butoxy-5-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Butoxy-5-fluorophenyl)boronic acid

A342858-100MG A342858-1G A342858-250MG A342858-5G
77263-784EA 20.6 USD
77263-784 77263-786 77263-788 77263-790
2-Butoxy-5-fluorophenylboronic acid 98%
2-Butoxy-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BFO₃
MW: 212.03 g/mol
MDL Number: MFCD05664232
CAS Number: 480438-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR