4-(trans-4-Ethylcyclohexyl)benzoic acid 98%

Supplier: Ambeed
Warning

A342708-1G A342708-25G
77278-000EA 33.99 USD
77278-000 77278-002 77278-004 77278-006
4-(trans-4-Ethylcyclohexyl)benzoic acid 98%
4-(trans-4-Ethylcyclohexyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₀O₂
MDL Number: MFCD06658176
CAS Number: 87592-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR