2,4-Dimethoxy-1-fluorobenzene 99%

Supplier: Ambeed
Warning

Synonyms: 1-Fluoro-2,4-dimethoxybenzene

A342603-1G A342603-25G A342603-5G
77251-418EA 23.69 USD
77251-418 77251-420 77251-422
2,4-Dimethoxy-1-fluorobenzene 99%
2,4-Dimethoxy-1-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₉FO₂
MW: 156.156 g/mol
MDL Number: MFCD00042265
CAS Number: 17715-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR