2-Bromo-1-(o-tolyl)ethanone 98%
Supplier: Ambeed
|
Danger
|
A342337-100MG
A342337-10G
A342337-1G
A342337-250MG
A342337-25G
A342337-5G
77263-764EA
7
USD
77263-764
77263-766
77263-768
77263-770
77263-772
77263-774
2-Bromo-1-(o-tolyl)ethanone 98%
2-Bromo-1-(o-tolyl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₉BrO |
MDL Number:
MFCD04038955 CAS Number: 51012-65-8 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...