5-Methoxy-1H-benzo[d][1,2,3]triazole 95%

Supplier: Ambeed
Warning

A342248-100MG A342248-1G A342248-250MG
77239-898EA 42 USD
77239-898 77239-900 77239-902
5-Methoxy-1H-benzo[d][1,2,3]triazole 95%
5-Methoxy-1H-benzo[d][1,2,3]triazole

Precaución: for Research Use Only

Formula: C₇H₇N₃O
MW: 149.15 g/mol
MDL Number: MFCD00962763
CAS Number: 27799-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR