Proparacaine hydrochloride 98+%

Supplier: Ambeed
Warning

Synonyms: 2-(Diethylamino)ethyl 3-Amino-4-propoxybenzoate Hydrochloride, Proxymetacaine Hydrochloride

A341969-100MG A341969-1G A341969-1MG A341969-250MG A341969-25MG A341969-5G
77263-752EA 14.42 USD
77263-752 77263-754 77263-756 77263-758 77263-760 77263-762
Proparacaine hydrochloride 98+%
Proparacaine hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₂₆N₂O₃
MW: 330.85 g/mol
MDL Number: MFCD00083467
CAS Number: 5875-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR