5-Bromo-4-methyl-1H-imidazole 95%

Supplier: Ambeed
Warning

A341968-10G A341968-1G A341968-250MG A341968-25G A341968-5G
77290-536EA 83 USD
77290-536 77290-538 77290-540 77290-542 77290-544
5-Bromo-4-methyl-1H-imidazole 95%
5-Bromo-4-methyl-1H-imidazole

Precaución: for Research Use Only

Formula: C₄H₅BrN₂
MDL Number: MFCD06659898
CAS Number: 15813-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR