2-Bromooctane 85%

Supplier: Ambeed
Warning

Synonyms: 2-Octyl bromide

A341902-1G A341902-5G A341902-25G
77263-750EA 17.51 USD
77263-750 77571-996 77746-669
2-Bromooctane 85%
2-Bromooctane

Precaución: for Research Use Only

Formula: H₃CBrCH(CH₂)₅CH₃
MW: 193.13 g/mol
MDL Number: MFCD00040823
CAS Number: 557-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR