4-(Methoxycarbonyl)picolinic acid 97%

Supplier: Ambeed
Warning

A341622-1G A341622-250MG A341622-5G A341622-100MG
77251-404EA 162.74 USD
77251-404 77251-406 77251-408 77746-668
4-(Methoxycarbonyl)picolinic acid 97%
4-(Methoxycarbonyl)picolinic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MW: 181.14 g/mol
MDL Number: MFCD01927895
CAS Number: 24195-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR