GSK-583 98%

Supplier: Ambeed
Warning

Synonyms: GSK583, 6-(tert-Butylsulphonyl)-N-(5-fluoro-1H-indazol-3-yl)quinolin-4-amine, 6‑(tert-Butylsulphonyl)‑N‑(5-fluoro‑1H‑indazol-3-yl)quinolin-4-amine

A341470-10MG A341470-1MG A341470-25MG A341470-50MG A341470-5MG A341470-2MG
77239-872EA 122.57 USD
77239-872 77239-874 77239-876 77239-878 77239-880 77616-502
GSK-583 98%
GSK-583

Precaución: for Research Use Only

Formula: C₂₀H₁₉FN₄O₂S
MW: 398.46 g/mol
MDL Number: MFCD30343846
CAS Number: 1346547-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR