tert-Butyl 2-hydroxycyclohexylcarbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (2-hydroxycyclohexyl)carbamate

A341416-1G A341416-250MG A341416-5G
77277-988EA 44.29 USD
77277-988 77277-990 77277-992
tert-Butyl 2-hydroxycyclohexylcarbamate 97%
tert-Butyl 2-hydroxycyclohexylcarbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
MW: 215.289 g/mol
MDL Number: MFCD08061947
CAS Number: 477584-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR