2-Octynoic acid 98%

Supplier: Ambeed
Danger

Synonyms: Oct-2-ynoic acid

A341312-1G A341312-25G A341312-5G
77277-978EA 19.57 USD
77277-978 77277-980 77277-982
2-Octynoic acid 98%
2-Octynoic acid

Precaución: for Research Use Only

Formula: C₈H₁₂O₂
MW: 140.18 g/mol
MDL Number: MFCD00004364
CAS Number: 5663-96-7
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR