2-Fluoro-6-(trifluoromethyl)phenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Fluoro-6-(trifluoromethyl)benzeneboronic acid
A341278-10G
A341278-1G
A341278-250MG
A341278-25G
A341278-5G
77277-968EA
114.33
USD
77277-968
77277-970
77277-972
77277-974
77277-976
2-Fluoro-6-(trifluoromethyl)phenylboronic acid 97%
2-Fluoro-6-(trifluoromethyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₅BF₄O₂ MW: 207.92 g/mol |
MDL Number:
MFCD07781239 CAS Number: 313545-34-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...