(S)-(-)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

Synonyms: (S)-Cl-MeO-BIPHEP

A341008-100MG A341008-250MG A341008-50MG A341008-1G
77251-388EA 98 USD
77251-388 77251-390 77251-392 77746-666
(S)-(-)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl 98%
(S)-(-)-5,5'-Dichloro-6,6'-dimethoxy-2,2'-bis(diphenylphosphino)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₃₈H₃₀Cl₂O₂P₂
MDL Number: MFCD03426987
CAS Number: 185913-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR