4-Bromo-2,5-xylidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2,5-dimethylaniline

A340690-100G A340690-1G A340690-25G A340690-5G
77290-520EA 198.79 USD
77290-520 77290-522 77290-524 77290-526
4-Bromo-2,5-xylidine 98%
4-Bromo-2,5-xylidine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.0757 g/mol
MDL Number: MFCD07780642
CAS Number: 30273-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR