2,3-Diamino-5-bromo-6-methylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5,6-Diamino-3-bromo-2-picoline, 5-Bromo-6-methylpyridine-2,3-diamine

A340675-10G A340675-1G A340675-250MG
77251-382EA 145.23 USD
77251-382 77251-384 77251-386
2,3-Diamino-5-bromo-6-methylpyridine 95%
2,3-Diamino-5-bromo-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₈BrN₃
MW: 202.055 g/mol
MDL Number: MFCD00160167
CAS Number: 59352-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR