5-Chloro-N-methylpyrazin-2-amine 97%

Supplier: Ambeed
Warning

A340372-100MG A340372-1G A340372-250MG
77263-736EA 80.34 USD
77263-736 77263-738 77263-740
5-Chloro-N-methylpyrazin-2-amine 97%
5-Chloro-N-methylpyrazin-2-amine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.57 g/mol
MDL Number: MFCD22398582
CAS Number: 1610667-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR