(6-Fluoro-2-pyridyl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-2-pyridinemethanol

A340272-100MG A340272-10G A340272-1G A340272-250MG
77263-728EA 15.45 USD
77263-728 77263-730 77263-732 77263-734
(6-Fluoro-2-pyridyl)methanol 97%
(6-Fluoro-2-pyridyl)methanol

Precaución: for Research Use Only

Formula: C₆H₆FNO
MW: 127.116 g/mol
MDL Number: MFCD07783827
CAS Number: 315180-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR