2-(4-Fluorophenyl)acetohydrazide 97%
Supplier: Ambeed
|
Warning
|
A340054-1G
A340054-5G
77290-516EA
19.57
USD
77290-516
77290-518
2-(4-Fluorophenyl)acetohydrazide 97%
2-(4-Fluorophenyl)acetohydrazide
Precaución: for Research Use Only
Formula:
C₈H₉FN₂O MW: 168.168 g/mol |
MDL Number:
MFCD06655053 CAS Number: 34547-28-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...