2-Chloroveratraldehyde 98%

Supplier: Ambeed
Warning

A340036-100G A340036-10G A340036-1G A340036-250MG A340036-25G A340036-5G
77277-950EA 139 USD
77277-950 77277-952 77277-954 77277-956 77277-958 77277-960
2-Chloroveratraldehyde 98%
2-Chloroveratraldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MDL Number: MFCD00084942
CAS Number: 5417-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR