3-Hydroxy-5-trifluoromethylthiophene-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-5-(trifluoromethyl)thiophene-2-carboxylic acid

A339914-100MG A339914-1G A339914-250MG A339914-5G
77277-942EA 54.59 USD
77277-942 77277-944 77277-946 77277-948
3-Hydroxy-5-trifluoromethylthiophene-2-carboxylic acid 97%
3-Hydroxy-5-trifluoromethylthiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃F₃O₃S
MW: 212.15 g/mol
MDL Number: MFCD05864365
CAS Number: 217959-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR