trans-3-(3-Pyridyl)acrylic acid 98%

Supplier: Ambeed
Warning

A339537-10G A339537-1G A339537-25G A339537-5G
77277-924EA 30.9 USD
77277-924 77277-926 77277-928 77277-930
trans-3-(3-Pyridyl)acrylic acid 98%
trans-3-(3-Pyridyl)acrylic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₂
MW: 149.15 g/mol
MDL Number: MFCD00006410
CAS Number: 19337-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR