2,2'-Bis[(4R)-4-Benzyl-2-Oxazoline] ≥97%, ee 99%

Supplier: Ambeed
Warning

A339529-100MG A339529-1G A339529-250MG A339529-5G
77263-720EA 66.95 USD
77263-720 77263-722 77263-724 77263-726
2,2'-Bis[(4R)-4-Benzyl-2-Oxazoline] ≥97%, ee 99%
2,2'-Bis[(4R)-4-Benzyl-2-Oxazoline]

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₂O₂
MW: 320.38 g/mol
CAS Number: 141362-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR