2,2'-Oxydiacetyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: Diglycolyl chloride, 2,2-Oxydiacetylchloride

A339483-5G A339483-25G A339483-1G
77251-376EA 33.99 USD
77251-376 77571-982 77746-659
2,2'-Oxydiacetyl chloride 97%
2,2'-Oxydiacetyl chloride

Precaución: for Research Use Only

Formula: C₄H₄Cl₂O₃
MW: 170.97 g/mol
MDL Number: MFCD00134471
CAS Number: 21062-20-4
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR