6,6'-Methylenebis(4-chloro-2-isopropyl-5-methylphenol) 97%

Supplier: Ambeed
Warning

A339396-100MG A339396-250MG
77290-512EA 77.25 USD
77290-512 77290-514
6,6'-Methylenebis(4-chloro-2-isopropyl-5-methylphenol) 97%
6,6'-Methylenebis(4-chloro-2-isopropyl-5-methylphenol)

Precaución: for Research Use Only

Formula: C₂₁H₂₆Cl₂O₂
MW: 381.34 g/mol
MDL Number: MFCD00865128
CAS Number: 15686-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR