4-Bromophenethyl bromide 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4-(2-bromoethyl)benzene

A338945-1G A338945-5G
77290-510EA 30 USD
77290-510 77550-974
4-Bromophenethyl bromide 98%
4-Bromophenethyl bromide

Precaución: for Research Use Only

Formula: C₈H₈Br₂
MW: 263.96 g/mol
MDL Number: MFCD00045052
CAS Number: 1746-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR