(E)-Methyl 3-(3-nitrophenyl)acrylate 98%

Supplier: Ambeed
Warning

A338846-100MG A338846-1G
77239-846EA 15.45 USD
77239-846 77239-848
(E)-Methyl 3-(3-nitrophenyl)acrylate 98%
(E)-Methyl 3-(3-nitrophenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₄
MW: 207.18 g/mol
MDL Number: MFCD00024570
CAS Number: 659-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR