(2-Trifluoromethylphenyl)methanesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: (2-(Trifluoromethyl)phenyl)methanesulphonamide

A338239-10G A338239-1G A338239-5G
77251-366EA 163.77 USD
77251-366 77251-368 77251-370
(2-Trifluoromethylphenyl)methanesulfonamide 98%
(2-Trifluoromethylphenyl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₈H₈F₃NO₂S
MW: 239.22 g/mol
MDL Number: MFCD09763601
CAS Number: 112941-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR