4-Carboxy-2-chlorophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Carboxy-2-chlorobenzeneboronic acid

A338181-10G A338181-1G A338181-5G
77277-920EA 42 USD
77277-920 77277-922 77616-492
4-Carboxy-2-chlorophenylboronic acid 97%
4-Carboxy-2-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BClO₄
MDL Number: MFCD07363763
CAS Number: 851335-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR