Trans-4-cyanocyclohexanecarboxylic acid 95%

Supplier: Ambeed
Warning

A337467-100MG A337467-1G A337467-250MG
77263-708EA 19 USD
77263-708 77263-710 77263-712
Trans-4-cyanocyclohexanecarboxylic acid 95%
Trans-4-cyanocyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MDL Number: MFCD27945176
CAS Number: 15177-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR