3,3''-Dibromo-1,1':3',1''-terphenyl 97%

Supplier: Ambeed
Warning

A337449-100MG A337449-1G A337449-250MG
77239-834EA 27 USD
77239-834 77239-836 77239-838
3,3''-Dibromo-1,1':3',1''-terphenyl 97%
3,3''-Dibromo-1,1':3',1''-terphenyl

Precaución: for Research Use Only

Formula: C₁₈H₁₂Br₂
MDL Number: MFCD28138089
CAS Number: 95962-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR