3,6-Pyridazinedicarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Pyridazine-3,6-dicarboxylic acid

A337282-100MG A337282-1G A337282-250MG
77277-908EA 36 USD
77277-908 77277-910 77277-912
3,6-Pyridazinedicarboxylic acid 98%
3,6-Pyridazinedicarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₄N₂O₄
MDL Number: MFCD01318693
CAS Number: 57266-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR