4'-Methylbutyrophenone 95%

Supplier: Ambeed
Warning

Synonyms: 1-(p-Tolyl)butan-1-one

A337258-100MG A337258-250MG
77290-502EA 46.35 USD
77290-502 77290-504
4'-Methylbutyrophenone 95%
4'-Methylbutyrophenone

Precaución: for Research Use Only

Formula: C₁₁H₁₄O
MW: 162.2283 g/mol
MDL Number: MFCD00027139
CAS Number: 4160-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR