(S)-Cyclobutyl(phenyl)methanamine hydrochloride 97%

Supplier: Ambeed
Warning

A337224-100MG A337224-1G A337224-250MG
77251-360EA 55.62 USD
77251-360 77251-362 77251-364
(S)-Cyclobutyl(phenyl)methanamine hydrochloride 97%
(S)-Cyclobutyl(phenyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅N·ClH
MW: 197.704 g/mol
MDL Number: MFCD20691543
CAS Number: 1202478-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR