2-Hydrazino-4-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydrazinyl-4-(trifluoromethyl)pyridine

A337149-250MG A337149-5G
77263-700EA 50.47 USD
77263-700 77263-702
2-Hydrazino-4-(trifluoromethyl)pyridine 98%
2-Hydrazino-4-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₆F₃N₃
MW: 177.13 g/mol
MDL Number: MFCD08056340
CAS Number: 89570-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR