(4-([1,1'-Biphenyl]-4-yl(phenyl)amino)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A337135-1G A337135-5G
77251-348EA 22 USD
77251-348 77251-350
(4-([1,1'-Biphenyl]-4-yl(phenyl)amino)phenyl)boronic acid 97%
(4-([1,1'-Biphenyl]-4-yl(phenyl)amino)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₂₄H₂₀BNO₂
MDL Number: MFCD28144504
CAS Number: 1084334-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR