1-Methyloxindole 98%

Supplier: Ambeed
Warning

A337076-1G A337076-250MG A337076-25G A337076-5G
77239-826EA 18 USD
77239-826 77239-828 77239-830 77239-832
1-Methyloxindole 98%
1-Methyloxindole

Precaución: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD00030253
CAS Number: 61-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR