2,6-Dimethoxy-p-benzoquinone 97%
Supplier: Ambeed
|
Warning
|
A336361-100MG
A336361-1MG
A336361-250MG
A336361-5G
A336361-1G
77251-328EA
7
USD
77251-328
77251-330
77251-332
77251-334
77616-484
2,6-Dimethoxy-p-benzoquinone 97%
2,6-Dimethoxy-p-benzoquinone
Precaución: for Research Use Only
Formula:
C₈H₈O₄ |
MDL Number:
MFCD00016368 CAS Number: 530-55-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...