N-Cbz-L-phenylalaninol 98%

Supplier: Ambeed
Warning

Synonyms: Z-Phenylalaninol, (S)-2-(Cbz-amino)-3-phenyl-1-propanol, N-Benzyloxycarbonyl-L-phenylalaninol

A335889-100G A335889-10G A335889-1G A335889-25G A335889-5G
77290-480EA 96.82 USD
77290-480 77290-482 77290-484 77290-486 77290-488
N-Cbz-L-phenylalaninol 98%
N-Cbz-L-phenylalaninol

Precaución: for Research Use Only

Formula: C₁₇H₁₉NO₃
MW: 285.3 g/mol
MDL Number: MFCD00191138
CAS Number: 6372-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR