2-(2-Chlorophenyl)-2-oxoacetic acid 95%

Supplier: Ambeed
Warning

A335637-100MG A335637-1G
77263-686EA 72.1 USD
77263-686 77263-688
2-(2-Chlorophenyl)-2-oxoacetic acid 95%
2-(2-Chlorophenyl)-2-oxoacetic acid

Precaución: for Research Use Only

Formula: C₈H₅ClO₃
MDL Number: MFCD06410833
CAS Number: 26118-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR