6-Bromo-3-chloroimidazo[1,2-a]pyridine 97%

Supplier: Ambeed
Warning

A335218-100MG A335218-1G A335218-250MG A335218-5G
77263-674EA 14 USD
77263-674 77263-676 77263-678 77263-680
6-Bromo-3-chloroimidazo[1,2-a]pyridine 97%
6-Bromo-3-chloroimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.48 g/mol
MDL Number: MFCD19105290
CAS Number: 1296224-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR