3-(1H-Imidazol-4-yl)propanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Deamino-histidine, Imidazole-5-propionic acid, Imidazolepropionic acid

A335215-250MG A335215-1G A335215-5G
77277-882EA 96.82 USD
77277-882 77616-474 77616-476
3-(1H-Imidazol-4-yl)propanoic acid 95%
3-(1H-Imidazol-4-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MW: 140.142 g/mol
MDL Number: MFCD00237415
CAS Number: 1074-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR