3-Bromoadamantane-1-carboxylic acid 98%

Supplier: Ambeed
Warning

A335161-1G A335161-25G A335161-5G
77277-876EA 5 USD
77277-876 77277-878 77277-880
3-Bromoadamantane-1-carboxylic acid 98%
3-Bromoadamantane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO₂
MDL Number: MFCD00167820
CAS Number: 21816-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR