3,6-Dibromo-9-vinyl-9H-carbazole 98%

Supplier: Ambeed
Warning

A335128-1G A335128-250MG
77277-872EA 53.56 USD
77277-872 77277-874
3,6-Dibromo-9-vinyl-9H-carbazole 98%
3,6-Dibromo-9-vinyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₄H₉Br₂N
MW: 351.03 g/mol
MDL Number: MFCD00080466
CAS Number: 1214-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR