4-(3-Bromophenyl)-1,3-thiazol-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: 4-(3-Bromophenyl)thiazol-2-amine, 2-Amino-4-(3-bromophenyl)thiazole

A334886-100MG A334886-250MG A334886-5G
77263-668EA 30.9 USD
77263-668 77263-670 77263-672
4-(3-Bromophenyl)-1,3-thiazol-2-amine 98%
4-(3-Bromophenyl)-1,3-thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₇BrN₂S
MW: 255.14 g/mol
MDL Number: MFCD00170238
CAS Number: 105512-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR